Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP tag Mouse anti-Tag, HRP, Clone: 1E10H7, Proteintech
SDP

Catalog No. 50553599 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-553-599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-553-599 Supplier Proteintech Group Inc Supplier No. HRP66002100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is conjugated with HRP. Note: Do not add azide (Sodium Azide or Smite) into the dilution buffer. azide is the HRP inhibitor which decreases the enzymatic activity of HRP.

The jellyfish Aequorea victoria contains green fluorescent protein (GFP) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressing and protein localization. Moreover, other applications for GFP include its use in assessing protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

Specifications

Antigen GFP tag
Applications Western Blot
Classification Monoclonal
Clone 1E10H7
Concentration 1.33 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.02% thimerosal; pH 7.3
Gene Alias CFP, eGFP, eYFP, GFP, GFP tag, HRP CFP, HRP eGFP, HRP GFP, HRP YFP, YFP
Host Species Mouse
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.