Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ GFP Transformation Extension: Colony PCR Experiment Kit

Shop All EDVOTEK Products


Click to view available options
:
GFP Transformation Extension: Colony PCR Experiment Kit
Description
Includes
Instructions, PCR EdvoBeads™, Colony Primer Mix Concentrate, Control DNA Concentrate, Ampicillin, IPTG, EdvoQuick™ DNA Ladder, TE Buffer, BactoBeads™, ReadyPour™ Luria Broth Agar, Petri Dishes, , ampicillin, IPTG, ReadyPour™ Luria Broth Agar (sterile), UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, InstaStain™ Ethidium Bromide, FlashBlue™ Liquid Stain, Petri Plates, Sterile Loops, Microcentrifuge & PCR Tubes & Wax Beads.
- Colony PCR to analyze bacteria transformed with pFluoroGreen
- Single colony used as DNA template for PCR
- Resulting PCR sample analyzed using agarose gel electrophoresis
- Used in conjunction Edvotek Transformation Kits (Part No: 222, 223 or 303)
Specifications
Specifications
Product Type | PCR Kit |
Grade | 9 to Undergraduate |
For Use With (Application) | Educational purpose |
Includes | Instructions, control BactoBeads™, ampicillin, IPTG, ReadyPour™ Luria Broth Agar (sterile), Petri Plates, sterile loops, PCR EdvoBeads™, control DNA and primers, microcentrifuge tubes, PCR tubes, agarose, DNA ladder, practice gel loading |
Time Range | 30 minutes (set up), 2 hours or overnight (PCR), 45 minutes (electrophoresis) |
Content And Storage | Refrigeration required |
Class Size | 10 Lab Groups |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction