Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFPT1 Antibody (SR2221), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322535100UL
Description
GFPT1 Monoclonal antibody specifically detects GFPT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
GFPT1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR2221 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, glutamine--fructose-6-phosphate transaminase 1, MSLG | |
A synthesized peptide derived from human GFPT1 (Uniprot #: Q06210) | |
100 μg | |
metabolism | |
2673 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction