Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFPT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576473 Supplier Invitrogen™ Supplier No. PA576473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFPT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GFPT1
Gene Accession No. P47856, P82808, Q06210
Gene Alias 2810423A18Rik; AI324119; AI449986; CMS12; CMSTA1; D-fructose-6-phosphate amidotransferase 1; eartha; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; Gfpt; GFPT1; GFPT1L; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine fructose-6-phosphate transaminase 1; glutamine:fructose 6 phosphate amidotransferase 1; glutamine:fructose-6-phosphate amidotransferase 1; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine--fructose-6-phosphate transaminase 1; harvest moon; hexosephosphate aminotransferase 1; hmn; MSLG; unm_z110
Gene Symbols GFPT1
Host Species Rabbit
Immunogen Recombinant fusion protein of human GFPT1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14583, 2673, 297417
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.