Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GFPT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109764
Description
Immunogen sequence: MCGIFAYLNY HVPRTRREIL ETLIKGLQRL EYRGYDSAGV GFDGGNDKDW EANACKIQLI KKKGKVKALD EEVHKQQDMD LDIEFDVHLG IAHTRWATHG EPSPVNSHPQ RSDKNNEFIV IHNGIITNYK DLKKFLESKG YDFESETDTE.
This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.
Specifications
| GFPT1 | |
| Polyclonal | |
| Unconjugated | |
| GFPT1 | |
| 2810423A18Rik; AI324119; AI449986; CMS12; CMSTA1; D-fructose-6-phosphate amidotransferase 1; eartha; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; Gfpt; GFPT1; GFPT1L; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine fructose-6-phosphate transaminase 1; glutamine:fructose 6 phosphate amidotransferase 1; glutamine:fructose-6-phosphate amidotransferase 1; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine--fructose-6-phosphate transaminase 1; harvest moon; hexosephosphate aminotransferase 1; hmn; MSLG; unm_z110 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14583, 2673, 297417 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.02 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P47856, P82808, Q06210 | |
| GFPT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GFPT1 (NP_002047.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction