Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304455100UL
Description
GFPT1 Polyclonal specifically detects GFPT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GFPT1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
2673 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, glutamine--fructose-6-phosphate transaminase 1, MSLG | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GFPT1 (NP_002047.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction