Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFPT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982819
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2819 Supplier Bioss Supplier No. BS13341R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate.

Specifications

Antigen GFPT1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GFPT1
Gene Accession No. Q06210
Gene Alias 2810423A18Rik; AI324119; AI449986; CMS12; CMSTA1; D-fructose-6-phosphate amidotransferase 1; eartha; GFA; GFAT; GFAT 1; GFAT1; GFAT1m; GFPT; Gfpt1; GFPT1L; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine fructose-6-phosphate transaminase 1; glutamine:fructose 6 phosphate amidotransferase 1; Glutamine:fructose-6-phosphate amidotransferase 1; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 1; glutamine--fructose-6-phosphate transaminase 1; harvest moon; Hexosephosphate aminotransferase 1; hmn; MSLG; unm_z110
Gene Symbols GFPT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GFPT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2673
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.