Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFPT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530340 Supplier Invitrogen™ Supplier No. PA530340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (97%), Rat (96%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFPT2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene GFPT2
Gene Accession No. O94808
Gene Alias AI480523; D-fructose-6-phosphate amidotransferase 2; GFAT; GFAT 2; GFAT2; GFPT2; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine fructose-6-phosphate transaminase 2; glutamine: fructose-6-phosphate aminotransferase 2; glutamine:fructose 6 phosphate amidotransferase 2; glutamine:fructose-6-phosphate amidotransferase 2; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine-fructose-6-phosphate transaminase 2; Hexosephosphate aminotransferase 2
Gene Symbols GFPT2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human GFPT2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9945
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.