Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GFPT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590419
Description
Immunogen sequence: RGSPLLIGVR SKYKLSTEQI PILYRTCTLE NVKNICKTRM KRLDSSACLH AVGDKAVEFF F; Positive Samples: 293T, HeLa, A375, NIH/3T3, Mouse heart, Mouse pancreas, Mouse testis, Rat heart.
GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.
Specifications
GFPT2 | |
Polyclonal | |
Unconjugated | |
GFPT2 | |
AI480523; D-fructose-6-phosphate amidotransferase 2; GFAT; GFAT 2; GFAT2; GFPT2; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine fructose-6-phosphate transaminase 2; glutamine: fructose-6-phosphate aminotransferase 2; glutamine:fructose 6 phosphate amidotransferase 2; glutamine:fructose-6-phosphate amidotransferase 2; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine-fructose-6-phosphate transaminase 2; Hexosephosphate aminotransferase 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14584, 360518, 9945 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
2.57 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O94808, Q4KMC4, Q9Z2Z9 | |
GFPT2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-260 of human GFPT2 (NP_0051011). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction