Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFPT2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 151891AP150UL

Description
GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.Specifications
GFPT2 | |
Polyclonal | |
Unconjugated | |
GFPT2 | |
GFAT 2, GFAT2, GFPT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9945 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O94808 | |
GFPT2 | |
GFPT2 Fusion Protein Ag7275 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction