Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-1/GDNF R alpha-1 Antibody (009), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290463JF549
Description
GFR alpha-1/GDNF R alpha-1 Monoclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GFR alpha-1/GDNF R alpha-1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
9 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse GFR alpha-1/GDNF R alpha-1 (AAB86600.1; Met 1-Ser 425). | |
0.1 mL | |
Neuroscience, Signal Transduction | |
2674 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction