Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFR alpha-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120909 Supplier Invitrogen™ Supplier No. PA5120909
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, SH-SY5Y, MCF7, Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, GPI-anchor, Lipid-anchor. Immunogen sequence: FGNGSDVTVW QPAFPVQTTT ATTTTALRVK NKPLGPAGSE NEIPTHVLPP CANLQAQKLK SNVSGNTHLC ISNGNY.

Glial cell line-derived neurotrophic factor (GDNF) is a potent survival factor for central and peripheral neurons and is essential for the development of kidneys and the enteric nerves system. Physiological responses to GDNF require the presence of a novel glycosylphosphadidylinositol linked protein GDNFRalpha, which is a cell surface receptor for GDNF. The cDNAs encoding GDNFRalpha from human, rat, chicken and mouse have been cloned recently. GDNFRalpha was also termed Ret ligand 1 (RETL1) or TGF-beta-related neurotrophic factor receptor 1 (TrnR1) and nominated as GFR-a-1 recently. GFR-a-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFR-a and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene GFRA1
Gene Accession No. P56159, P97785, Q62997
Gene Alias AU042498; DKFZp313E1012; DKFZp686J0156; FLJ10561; FLJ31546; GDNF family receptor alpha 1; GDNF family receptor alpha-1; GDNF receptor alpha; GDNF receptor alpha-1; GDNFR; GDNFRA; GDNFR-alpha; GDNFR-alpha-1; gfr alpha 1; GFR alpha-1; GFRA1; GFR-alpha-1; glial cell line derived neurotrophic factor family receptor alpha 1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; MGC23045; PI-linked cell-surface accessory protein; RET ligand 1; RET1L; RETL1; TGF-beta-related neurotrophic factor receptor 1; TRNR1
Gene Symbols GFRA1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14585, 25454, 2674
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.