Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA132476
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA132476 100 μg
1 options

Catalog No. PIPA132476

Supplier: Thermo Scientific PA132476

Rabbit Polyclonal Antibody

PA1-32476 detects GFR alpha 1 from human, rat, mouse samples. PA1-32476 has been successfully used in Western blot applications. The PA1-32476 immunogen is: Synthetic peptide: TALRVKNKPLGPAG, corresponding to amino acids 369-382 of Human GFR alpha 1.

Glial cell line-derived neurotrophic factor (GDNF) and neurturin (NTN) are two structurally related, potent neurotrophic factors that play key roles in the control of neuron survival and differentiation. The protein encoded by this gene is a member of the GDNF receptor family. It is a glycosylphosphatidylinositol(GPI)-linked cell surface receptor for both GDNF and NTN, and mediates activation of the RET tyrosine kinase receptor. This gene is a candidate gene for Hirschsprung disease. Multiple alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Gene GFRA1
Gene Accession No. P56159, P97785, Q62997
Gene Alias DKFZp313E1012, DKFZp686J0156, FLJ10561, FLJ31546, GDNFR, GDNFRA, GFR-ALPHA-1, MGC23045, RET1L, RETL1, TRNR1
Gene Symbols GFRA1
Host Species Rabbit
Immunogen synthetic peptide TALRVKNKPLGPAG, corresponding to amino acids 369-382 of Human GFR alpha 1.
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14585, 25454, 2674
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.