Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-2/GDNF R alpha-2 Antibody (101), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289419MFV450
Description
GFR alpha-2/GDNF R alpha-2 Monoclonal antibody specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for ELISASpecifications
GFR alpha-2/GDNF R alpha-2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
101 | |
ELISA | |
GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human GFR alpha-2/GDNF R alpha-2 (Accession#: NP_001486.4; Met 1-Ser 441). | |
0.1 mL | |
Cytokine Research, Immunology | |
2675 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction