Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-2/GDNF R alpha-2 Rabbit anti-Human, Clone: 104, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | GFR alpha-2/GDNF R alpha-2 |
---|---|
Clone | 104 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
GFR alpha-2/GDNF R alpha-2 Monoclonal specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Western Blot.Specifications
GFR alpha-2/GDNF R alpha-2 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA | |
Recombinant Human GFR alpha-2/GDNF R alpha-2 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
104 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology | |
0.2 um filtered solution in PBS | |
2675 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title