Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFR alpha-2 Recombinant Rabbit Monoclonal Antibody (101), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543099
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543099 100 μL
PIMA543448 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543099 Supplier Invitrogen™ Supplier No. MA543099
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human GFRA2

GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-2
Applications ELISA
Classification Recombinant Monoclonal
Clone 101
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GFRA2
Gene Accession No. O00451
Gene Alias GDNF family receptor alpha 2; GDNF family receptor alpha-2; GDNF re; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; Gdnfrb; GDNFR-beta; Gfra2; GFR-alpha 2; GFR-alpha-2; glial cell line derived neurotrophic factor family receptor alpha 2; glial cell line derived neurotrophic factor receptor, beta; HpV18gp1; neurturin receptor alpha; NRTNR-alpha; NTNRA; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; RETL2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored; TRNR2; tyrosine kinase receptor ligand 2
Gene Symbols GFRA2
Host Species Rabbit
Immunogen Recombinant Human GFRA2 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2675
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.