Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFR alpha-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582733 Supplier Invitrogen™ Supplier No. PA582733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPTESRLMNS CLQARRKCQA DPTCSAAYHH LDSCTSSIST PLPSEEPSVP ADCLEAAQQL RNSSLIGCMC HRRMKNQVAC L Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 69%.

GFRa-3 binds persephin, thus, persephin, GFRa-3, and Ret PTK form a complex to transduce persephin signal and to mediate persephin function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gfra3
Gene Accession No. O60609
Gene Alias GDNF family receptor alpha 3; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNF-family receptor alpha 3; GDNFR3; GDNFR-alpha-3; GFR alpha-3; GFRA3; GFRalpha3; GFR-alpha-3; glial cell line derived neurotrophic factor family receptor alpha 3; glial cell line-derived neurotrophic factor receptor alpha-3; GPI-linked receptor; UNQ339/PRO538/PRO3664; Y15110
Gene Symbols Gfra3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GFRA3. Recombinant protein control fragment (Product #RP-92873).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2676
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.