Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-4/GDNF R alpha-4/Persephin receptor Antibody (MM0311-3P34) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | GFR alpha-4/GDNF R alpha-4/Persephin receptor |
---|---|
Clone | MM0311-3P34 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Human samples. It is validated for Western Blot.Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
NP_071422 | |
64096 | |
IgG1 | |
Protein G purified |
MM0311-3P34 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR receptor alpha 4, GFR-alpha-4, Persephin receptor | |
GFRA4 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title