Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFR alpha-4/GDNF R alpha-4/Persephin receptor Antibody (MM0311-3P34), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212414IR
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Human samples. It is validated for Western Blot.Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GFRA4 | |
Human recombinant GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
0.1 mL | |
64096 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
MM0311-3P34 | |
Western Blot | |
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR receptor alpha 4, GFR-alpha-4, Persephin receptor | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction