Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFR alpha-4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA547182
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547182 Supplier Thermo Scientific Supplier No. PA547182

Goat Polyclonal Antibody

In direct ELISAs, approximately 15% cross-reactivity with recombinant human GFR alpha -4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

GFR alpha-4 Polyclonal antibody specifically detects GFR alpha-4 in Mouse samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-4
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Gfra4
Gene Accession No. Q9JJT2
Gene Alias AW060733; G630015H18Rik; GDNF family receptor alpha 4; GDNF family receptor alpha-4; GDNF R alpha-4; GDNF receptor alpha-4; GDNFR-alpha-4; GFR alpha-4; GFR receptor alpha 4; GFRA4; GFR-alpha-4; glial cell line derived neurotrophic factor family receptor alpha 4; neurotrophic factor receptor splice variant A (Gfra4 gene); persephin receptor
Gene Symbols Gfra4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 (aa 35-242)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14588
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.