Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFRA4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980111
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0111 Supplier Bioss Supplier No. BS0168R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the GDNF receptor family. It is a glycosylphosphatidylinositol(GPI)-linked cell surface receptor for persephin, and mediates activation of the RET tyrosine kinase receptor. This gene is a candidate gene for RET-associated diseases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Jul 2008].

Specifications

Antigen GFRA4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gfra4
Gene Accession No. Q9GZZ7
Gene Alias AW060733; G630015H18Rik; GDNF family receptor alpha 4; GDNF family receptor alpha-4; GDNF R alpha-4; GDNF receptor alpha-4; GDNFR-alpha-4; GFR alpha-4; GFR receptor alpha 4; Gfra4; GFR-alpha-4; glial cell line derived neurotrophic factor family receptor alpha 4; neurotrophic factor receptor splice variant A (Gfra4 gene); Persephin receptor
Gene Symbols Gfra4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GFRA4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64096
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.