Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFRAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112099 Supplier Invitrogen™ Supplier No. PA5112099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GFRAL Polyclonal Antibody for Western Blot

Brainstem-restricted receptor for GDF15 which regulates food intake, energy expenditure and body weight in response to metabolic and toxin-induced stresses (PubMed:28953886, PubMed:28846097, PubMed:28846098, PubMed:28846099). Upon interaction with its ligand, GDF15, interacts with RET and induces cellular signaling through activation of MAPK- and AKT- signaling pathways. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFRAL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GFRAL
Gene Accession No. Q6UXV0
Gene Alias AY457637; bA360D14.1; C6orf144; GDNF family receptor alpha like; GDNF family receptor alpha-like; GDNF receptor alpha-like; GFRAL; Gral; IVFI9356; RGD1565220; UNQ9356; UNQ9356/PRO34128
Gene Symbols GFRAL
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Human GFRAL.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.