Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561699 Supplier Invitrogen™ Supplier No. PA561699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSMESRPPAQ TSLPASSGLD DLDLLGKTLL QQSLPPESQQ VRWEKQQPTP RLTLRDLQNK SSS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

This gene encodes an apoptosis inhibitory protein whose expression prevents apoptosis after growth factor deprivation. This protein suppresses the transcription factor E2F1-induced apoptosis and also interacts with, and negatively regulates Acinus, a nuclear factor involved in apoptotic DNA fragmentation. Its depletion enhances the cytotoxic action of the chemotherapeutic drugs. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GGA1
Gene Accession No. Q9UJY5
Gene Alias 4930406E12Rik; ADP-ribosylation factor binding protein 1; ADP-ribosylation factor-binding protein GGA1; AU016030; AW209092; gamma-adaptin related protein 1; gamma-adaptin-related protein 1; GGA1; golgi associated, gamma adaptin ear containing, ARF binding protein 1; golgi-associated, gamma adaptin ear containing, ARF binding protein 1; golgi-localized, gamma ear-containing, ARF-binding protein 1
Gene Symbols GGA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GGA1. Recombinant protein control fragment (Product #RP-100901).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26088
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.