Learn More
GGA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305368AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) protein family. Members of this family are ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
GGA1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UJY5 | |
GGA1 | |
Between 425 and 475 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GGA1 | |
ADP-ribosylation factor binding protein GGA1, Golgi-localized, gamma ear-containing, ARF-binding protein 1, Gamma-adaptin related protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26088 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.