Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GGA2 Antibody (OTI1D2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271725AF350
Description
GGA2 Monoclonal specifically detects GGA2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GGA2 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
GGA2 | |
Full length recombinant protein of human GGA2 (NP_055859) produced in HEK293T cells | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI1D2 | |
Western Blot | |
FLJ20966, Gamma-adaptin-related protein 2, golgi associated, gamma adaptin ear containing, ARF binding protein 2, golgi-associated, gamma adaptin ear containing, ARF binding protein 2, Golgi-localized, gamma ear-containing, ARF-binding protein 2, KIAA1080ADP-ribosylation factor-binding protein GGA2, VEAR, VHS domain and ear domain of gamma-adaptin, VHS domain and ear domain-containing protein | |
Mouse | |
0.1 mL | |
23062 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction