Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545523 Supplier Invitrogen™ Supplier No. PA545523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MLKKQGIIKQ DPKLPVDKIL PPPSPWPKSS IFDADEEKSK LLTRLLKSNH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GGA2
Gene Accession No. Q9UJY4
Gene Alias 1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-localized, gamma ear-containing, ARF-binding protein 2; Kiaa1080; mKIAA1080; VEAR; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein
Gene Symbols GGA2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GGA2 (aa 154-203).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23062
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.