Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588267 Supplier Invitrogen™ Supplier No. PA588267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAATAVAAAV AGTESAQGPP GPAASLELWL NKATDPSMSE QDWSAIQNFC EQVNTDPNGP THAPWLLAHK IQSPQEKEAL YALTVLEMCM NHCGEKFHSE VAKFRFLNEL IKVLSPKYLG SWATGKVKGR VIEILFSWTV WFPEDIKIRD AYQMLKKQGI IKQDPKLPVD KILPPPSPWP KSSIFDADEE KSKLLTRLLK SNHPEDLQAA NRLIKNLVKE EQEKSEKVSK RVSAVEEVRS HVKVLQEMLS MYRRPGQAPP DQEALQVVYE RCEKLRPTLF RLASDTTDDD DALAEILQAN; Positive Samples: HepG2, Raji, MCF7, THP-1, 22Rv1, Mouse brain, Mouse kidney; Cellular Location: Endosome membrane, Golgi apparatus, Peripheral membrane protein, trans-Golgi network membrane.

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. This family member may play a significant role in cargo molecules regulation and clathrin-coated vesicle assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGA2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GGA2
Gene Accession No. Q6P5E6, Q9UJY4
Gene Alias 1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-localized, gamma ear-containing, ARF-binding protein 2; Kiaa1080; mKIAA1080; VEAR; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein
Gene Symbols GGA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GGA2 (NP_0558591).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23062, 74105
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.