Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528462
Description
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants have been identified in this gene.
Specifications
| GGA3 | |
| Polyclonal | |
| Unconjugated | |
| GGA3 | |
| ADP-ribosylation factor binding protein 3; ADP-ribosylation factor-binding protein GGA3; ADP-ribosylation factor-binding protein GGA3a; C230037M19Rik; Gga3; gga3a; ggolgi-associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3a; golgi-localized, gamma ear-containing, ARF-binding protein 3; im:7144559; KIAA0154; mKIAA0154; si:ch211-108p22.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23163, 260302 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.59 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q8BMI3, Q9NZ52 | |
| GGA3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GGA3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction