Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582888 Supplier Invitrogen™ Supplier No. PA582888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HHLDALDQLL EEAKVTSGLV KPTTSPLIPT TTPARPLLPF STGPGSPLFQ PLSFQSQGSP PKGPELSLAS IHVPLESIKP SSALPVTAYD K Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 82%.

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGA3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GGA3
Gene Accession No. Q9NZ52
Gene Alias ADP-ribosylation factor binding protein 3; ADP-ribosylation factor-binding protein GGA3; ADP-ribosylation factor-binding protein GGA3a; C230037M19Rik; Gga3; gga3a; ggolgi-associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3a; golgi-localized, gamma ear-containing, ARF-binding protein 3; im:7144559; KIAA0154; mKIAA0154; si:ch211-108p22.4
Gene Symbols GGA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GGA3. Recombinant protein control fragment (Product #RP-104576).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23163
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.