Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117967 Supplier Invitrogen™ Supplier No. PA5117967
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GGA3 Polyclonal Antibody for Western Blot, ICC/IF, IP

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGA3
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene GGA3
Gene Accession No. Q9NZ52
Gene Alias ADP-ribosylation factor binding protein 3; ADP-ribosylation factor-binding protein GGA3; ADP-ribosylation factor-binding protein GGA3a; C230037M19Rik; Gga3; gga3a; ggolgi-associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3a; golgi-localized, gamma ear-containing, ARF-binding protein 3; im:7144559; KIAA0154; mKIAA0154; si:ch211-108p22.4
Gene Symbols GGA3
Host Species Rabbit
Immunogen E. coli-derived Human GGA3 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23163
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.