Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGA3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982822
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2822 Supplier Bioss Supplier No. BS13345R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants have been identified in this gene.

Specifications

Antigen GGA3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GGA3
Gene Accession No. Q9NZ52
Gene Alias ADP-ribosylation factor binding protein 3; ADP-ribosylation factor-binding protein GGA3; BOS_19551; C230037M19Rik; Gga3; gga3a; ggolgi-associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3a; golgi-associated, gamma adaptin ear containing, ARF binding protein 3; Golgi-localized, gamma ear-containing, ARF-binding protein 3; im:7144559; KIAA0154; mKIAA0154; si:ch211-108p22.4
Gene Symbols GGA3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GGA3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23163
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.