Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGA3Rabbit anti-Chimpanzee, Human, Polyclonal, Rockland™
SDP

Catalog No. 501991846
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1846 Supplier Rockland Immunochemicals Supplier No. 600401893
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human GGA3 protein. A BLAST analysis was used to suggest reactivity with this protein from human and chimpanzee based on 100% homology for the immunogen sequence. Cross reactivity may occur with GGA3 from macaque (93% homology) and orangutan (87% homology) sources. The immunizing sequence is present on both the long (isoform 1) and short (isoform 2) transcript variants for this protein.

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants have been identified in this gene.

Specifications

Antigen GGA3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene GGA3
Gene Accession No. Q9NZ52
Gene Alias ADP-ribosylation factor binding protein 3; ADP-ribosylation factor-binding protein GGA3; BOS_19551; C230037M19Rik; Gga3; gga3a; ggolgi-associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3; golgi associated, gamma adaptin ear containing, ARF binding protein 3a; golgi-associated, gamma adaptin ear containing, ARF binding protein 3; Golgi-localized, gamma ear-containing, ARF-binding protein 3; im:7144559; KIAA0154; mKIAA0154; si:ch211-108p22.4
Gene Symbols GGA3
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23163, 454877
Target Species Chimpanzee, Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.