Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGCT Monoclonal Antibody (04)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530460
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human GGCT.
GGCT (EC 2.3.2.4) catalyzes the formation of 5-oxoproline (pyroglutamic acid) from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis.
Specifications
GGCT | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O75223 | |
GGCT | |
Recombinant Human GGCT Protein (Met1-Leu188). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
04 | |
Unconjugated | |
GGCT | |
A030007L17Rik; AA673177; C7orf24; CRF21; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; gamma-glutamylcyclotransferase; Gctg; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590 | |
Mouse | |
Protein A | |
RUO | |
79017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction