Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGCT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556295 Supplier Invitrogen™ Supplier No. PA556295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSGCKDVTGP DEESFLYFAY GSNLLTERIH LRNPSAAFFC VARLQDFKLD FGNSQGKTSQ TWHGGIATIF QSPGDEVWGV VWKMNKSNLN SLDE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.

GGCT (EC 2.3.2.4) catalyzes the formation of 5-oxoproline (pyroglutamic acid) from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGCT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GGCT
Gene Accession No. O75223
Gene Alias A030007L17Rik; AA673177; C7orf24; CRF21; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; gamma-glutamylcyclotransferase; Gctg; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590
Gene Symbols GGCT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GGCT. Recombinant protein control fragment (Product #RP-92598).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.