Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGCT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115284 Supplier Invitrogen™ Supplier No. PA5115284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GGCT.

GGCT (EC 2.3.2.4) catalyzes the formation of 5-oxoproline (pyroglutamic acid) from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGCT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GGCT
Gene Accession No. O75223, Q9D7X8
Gene Alias A030007L17Rik; AA673177; C7orf24; CRF21; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; gamma-glutamylcyclotransferase; Gctg; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590
Gene Symbols GGCT
Host Species Rabbit
Immunogen A synthesized peptide derived from human GGCT(Accession O75223), corresponding to amino acid residues E109-K159.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110175, 362368, 79017
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.