Learn More
GGCT Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305563A

Description
The recommended shelf life for this product is 1 year from date of receipt.
GGCT (EC 2.3.2.4) catalyzes the formation of 5-oxoproline (pyroglutamic acid) from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis.Specifications
GGCT | |
Polyclonal | |
Unconjugated | |
GGCT | |
C7orf24, CRF21, GCTG, GGC, cytochrome c-releasing factor 21, gamma -glutamyl cyclotransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79017 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O75223 | |
GGCT | |
Between 138 and 188 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.