Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GGCX Polyclonal Antibody
GGCX Polyclonal Antibody
Supplier: Thermo Scientific PA550417
Description
GGCX Polyclonal Antibody for Western Blot, IHC (P)

Specifications
GGCX | |
Polyclonal | |
Unconjugated | |
Ggcx | |
FLJ26629; Gamma-glutamyl carboxylase; GC; GGCX; Peptidyl-glutamate 4-carboxylase; vitamin K gamma glutamyl carboxylase; Vitamin K-dependent gamma-carboxylase; VKCFD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2677 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P38435 | |
Ggcx | |
Fusion protein corresponding to residues near the C terminal of human gamma-glutamyl carboxylase. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction