Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596332 Supplier Invitrogen™ Supplier No. PA596332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPHGDTAKKP IIGILMQKCR NKVMKNYGRY YIAASYVKYL ESAGARVVPV RLDLTEKDYE ILFKSINGIL FPGGSVDLRR SDYAKVAKIF YNLSIQSFDD GDYFPVWGTC LGFEELSLLI SGECLLTATD TVDVAMPLNF TGGQLHSRMF QNFPTELLLS LAVEPLTANF HKWSLSVKNF TMNEKLKKFF NVLTTNTDGK IEFIST; Positive Samples: HeLa, Mouse kidney; Cellular Location: Lysosome, Melanosome, extracellular space.

Gamma-Glutamyl hydrolase (GGH) is a homodimeric protein that catalyzes the cleavage of the gamma-glutamyl chain of folylpoly-gamma-glutamyl substrates and is a central enzyme in folyl and antifolyl poly-gamma-glutamate metabolism. The activities of the baculovirus-expressed enzyme and the enzyme from HL-60 cells were similar on methotrexate polyglutamates. GGH is expressed in a variety of human tumors and correlates with poor prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ggh
Gene Accession No. Q92820, Q9Z0L8
Gene Alias conjugase; FGPH; folylpolyglutamate hydrolase; gamma-GH; gamma-glutamyl hydrolase; gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase); Gamma-Glu-X carboxypeptidase; GGH; GH
Gene Symbols Ggh
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-230 of human GGH (NP_0038691).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14590, 8836
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.