Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGNBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570728 Supplier Invitrogen™ Supplier No. PA570728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Horse: 79%; Human: 100%; Rat: 79%.

This gene is the ortholog of the mouse gametogenetin-binding protein 1 gene. In human, the open reading frame is disrupted by a nonsense mutation after 8-aa; consequently, this gene is currently considered to be a unitary pseudogene in human even though it is functional in other mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGNBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GGNBP1
Gene Accession No. Q5YKI7
Gene Alias gametogenetin binding protein 1; gametogenetin binding protein 1 (pseudogene); gametogenetin-binding protein 1; Ggnbp1
Gene Symbols GGNBP1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human GGNBP1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 449520
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.