Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGNBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567289 Supplier Invitrogen™ Supplier No. PA567289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEEEFPFERR QIPLYIDDTL TMVMEFPDNV LNLDGHQNNG AQLKQFIQRH GMLKQQDLSI AMVVTSREVL SALSQLVPCV GCRRSVERLF SQLV Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

May be involved in spermatogenesis. Its mouse homolog is induced by dioxin and may play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGNBP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GGNBP2
Gene Accession No. Q9H3C7
Gene Alias AI451896; C3HC4-type zinc finger protein; C75939; D330017P12Rik; DIF3; DIF-3; dioxin inducible factor 3; dioxin-inducible factor 3; gametogenetin binding protein 2; gametogenetin-binding protein 2; gametogenin binding protein 2; Ggnbp2; laryngeal carcinoma related gene 1; laryngeal carcinoma-related protein 1; LCRG1; LZK1; Protein ZNF403; Zfp403; zinc finger protein 403; ZNF403
Gene Symbols GGNBP2
Host Species Rabbit
Immunogen Recombinant Human GGNBP2. Recombinant protein control fragment (Product #RP-107927).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.