Learn More
Invitrogen™ GGNBP2 Polyclonal Antibody

Description
This antibody is predicted to react with chicken, mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | GGNBP2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GGNBP2 |
| Gene Accession No. | Q9H3C7 |
| Gene Alias | AI451896; C3HC4-type zinc finger protein; C75939; D330017P12Rik; DIF3; DIF-3; dioxin inducible factor 3; dioxin-inducible factor 3; gametogenetin binding protein 2; gametogenetin-binding protein 2; gametogenin binding protein 2; Ggnbp2; laryngeal carcinoma related gene 1; laryngeal carcinoma-related protein 1; LCRG1; LZK1; Protein ZNF403; Zfp403; zinc finger protein 403; ZNF403 |
| Gene Symbols | GGNBP2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.