Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGPS1 Monoclonal Antibody (OTI2E7), TrueMAB™, OriGene
SDP

Catalog No. 502542063
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2063 30 μL
50-167-2847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2063 Supplier OriGene Supplier No. TA501729S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GGPS1 Monoclonal antibody specifically detects GGPS1 in Human samples. It is validated for Western Blot

Specifications

Antigen GGPS1
Applications Western Blot
Classification Monoclonal
Clone OTI2E7
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SMAD2
Gene Accession No. Q15796
Gene Alias (2E,6E)-farnesyl diphosphate synthase; 1810026C22Rik; 9530089B04Rik; AI843169; C79210; Crlf3; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyltranstransferase; Geranylgeranyl diphosphate synthase; geranylgeranyl diphosphate synthase 1; geranylgeranyl pyrophosphate synthase; geranylgeranyl pyrophosphate synthetase; geranyltranstransferase; GGPP synthase; GGPP synthetase; GGPPS; GGPPS1; GGPPSase; GGPS1; rGGPS1a; rGGPS1a1; rGGPS1a2; rGGPS1a3
Host Species Mouse
Immunogen Full length human recombinant protein of human SMAD2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4087
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.