Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583091 Supplier Invitrogen™ Supplier No. PA583091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CEDLTEGKFS FPTIHAIWSR PESTQVQNIL RQRTENIDIK KYCVHYLEDV GSFEYTRNTL KELEAKAYKQ IDARGGNPEL VALVKHLSKM FKE Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 91%.

This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGPS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GGPS1
Gene Accession No. O95749
Gene Alias (2E,6E)-farnesyl diphosphate synthase; 1810026C22Rik; 9530089B04Rik; AI843169; C79210; Crlf3; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyltranstransferase; Geranylgeranyl diphosphate synthase; geranylgeranyl diphosphate synthase 1; geranylgeranyl pyrophosphate synthase; geranylgeranyl pyrophosphate synthetase; geranyltranstransferase; GGPP synthase; GGPP synthetase; GGPPS; GGPPS1; GGPPSase; GGPS1; rGGPS1a; rGGPS1a1; rGGPS1a2; rGGPS1a3
Gene Symbols GGPS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GGPS1. Recombinant protein control fragment (Product #RP-91749).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9453
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.