Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GGT1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298438APC
Description
GGT1 Polyclonal antibody specifically detects GGT1 in Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
GGT1 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
CD224, CD224 antigen, D22S672, D22S732, EC 2.3.2.2, gamma-glutamyl transpeptidase, gamma-glutamyltransferase 1MGC96904, gamma-glutamyltranspeptidase 1, GGT 1, GGTMGC96963, glutamyl transpeptidase, GTG, MGC96892 | |
Produced in rabbits immunized with purified, recombinant Rat GGT1 (Uniprot#: P07314; Pro27-Tyr568). | |
0.1 mL | |
Cancer | |
2678 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction