Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GGT1 Monoclonal Antibody (138H11), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA511815
Description
MA5-11815 targets Glutamyl Transferase gamma in FACS, IHC (F), IP, and WB applications and shows reactivity with Human samples. The MA5-11815 immunogen is purified human kidney GGT.
Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.
Specifications
GGT1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P19440 | |
GGT1 | |
Purified human kidney GGT | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
138H11 | |
Unconjugated | |
GGT1 | |
CD224; D22S672; D22S732; dwg; gamma glutamyl-transferase 1; gamma-glutamyl transpeptidase; gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGLUT; Ggt; GGT 1; Ggt1; GGT-1; Ggtp; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; GTG; Leukotriene-C4 hydrolase; testicular tissue protein Li 73 | |
Mouse | |
Protein G | |
RUO | |
2678 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction