Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGT1 Monoclonal Antibody (1F1B5)
GREENER_CHOICE

Catalog No. FERMA548466
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548466 Supplier Thermo Scientific Supplier No. MA548466
Add to Cart
Add to Cart

GGT1 Monoclonal Antibody (1F1B5)

GGT1 Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen GGT1
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1F1B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GGT1
Gene Accession No. P19440
Gene Alias CD224; D22S672; D22S732; dwg; gamma glutamyl-transferase 1; gamma-glutamyl transpeptidase; gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGLUT; Ggt; GGT 1; Ggt1; GGT-1; Ggtp; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; GTG; Leukotriene-C4 hydrolase; testicular tissue protein Li 73
Gene Symbols GGT1
Host Species Mouse
Immunogen Purified recombinant fragment of human CD224 (AA: extra 381-470) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.