Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGT1 Mouse anti-Human, Mouse, Unconjugated, Clone: 1F9, Abnova™

Catalog No. 89339158 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-158 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-158 Supplier Abnova Corporation Supplier No. H00002678M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GGT1 Monoclonal antibody specifically detects GGT1 in Human, Mouse samples. It is validated for WB-Ce, IHC-P, S-ELISA, ELISA, WB-Re, IP.

Specifications

Antigen GGT1
Applications Immunohistochemistry (Paraffin), ELISA, Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1F9
Conjugate Unconjugated
Description gamma-glutamyltransferase 1
Gene GGT1
Gene Accession No. NM_005265.2
Gene Alias CD224, D22S672, D22S732, GGT, GTG, MGC96892, MGC96904, MGC96963
Host Species Mouse
Immunogen GGT1 (NP_005256.2, 381 a.a. ∼ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 2678
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.