Learn More
Invitrogen™ GGT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120579
Description
Positive test controls include: Jurkat, Mouse kidney. The target is usually found in the following locations: Cell membrane, Single-pass type II membrane protein. Immunogen sequence: KPEFYTPDDG GTAHLSVVAE DGSAVSATST INLYFGSKVR SPVSGILFNN EMDDFSSPSI TNEFGVPPSP ANFIQPGKQP LSSMCPTIMV GQDGQVRMVV GAAGGTQITT ATALAIIYNL WFGYDVKRAV EEPRLHNQLL PNVTTVERNI DQAVTAALET RHHHTQIAST FIAVVQAIVR TAGGWAAASD SRKGGEPAGY.
Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.
Specifications
GGT1 | |
Polyclonal | |
Unconjugated | |
GGT1 | |
CD224; D22S672; D22S732; dwg; gamma glutamyl-transferase 1; gamma-glutamyl transpeptidase; gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGLUT; Ggt; GGT 1; Ggt1; GGT-1; Ggtp; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; GTG; Leukotriene-C4 hydrolase; testicular tissue protein Li 73 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116568, 14598, 2678 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07314, P19440, Q60928 | |
GGT1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 370-569 of human GGT1 (NP_0382652?). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.