Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGT3P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572348
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572348 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572348 Supplier Invitrogen™ Supplier No. PA572348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GGT3P Polyclonal Antibody for Western Blot

Initiates extracellular glutathione (GSH) breakdown; catalyzes the transfer of the glutamyl moiety of glutathione to amino acids and dipeptide acceptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGT3P
Applications Western Blot
Classification Polyclonal
Concentration 2.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GGT3P
Gene Accession No. A6NGU5
Gene Alias gamma-glutamyltransferase 3 pseudogene; GGT3; GGT3P
Gene Symbols GGT3P
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 176-207 amino acids from the Central region of human GGT3P.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2679
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.