Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112164 Supplier Invitrogen™ Supplier No. PA5112164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product can be used in direct ELISA.

This gene is a member of a gene family that encodes gamma-glutamyl transpeptidase enzymes. This enzyme consists of a heavy and a light chain, and is able to hydrolyze the gamma-glutamyl moiety of glutathione. It converts leukotriene C4 to leukotriene D4, however, it doesn't convert synthetic substrates that are commonly used to assay gamma-glutamyl transpeptidase. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGT5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5.35 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GGT5
Gene Accession No. P36269
Gene Alias AI551243; gamma-glutamyl cleaving enzyme; gamma-glutamyl leukotrienase; Gamma-glutamyl transpeptidase-related enzyme; gamma-glutamyl transpeptidase-related protein; gamma-glutamyltransferase 5; Gamma-glutamyltransferase-like activity 1; gamma-glutamyltranspeptidase 5; GGL; GGT 5; GGT5; Ggta1; GGTLA1; GGT-rel; glutathione hydrolase 5; Glutathione hydrolase 5 heavy chain; Glutathione hydrolase 5 light chain; Glutathione hydrolase 5 proenzyme; Leukotriene-C4 hydrolase
Gene Symbols GGT5
Host Species Rabbit
Immunogen Recombinant Human GGT5/GGTLA1 Protein (30-586).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2687
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.